siRNA Details
Gene Symbol: Srsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109552.2 | NP_001103022.1 | 1 | 230 | ||
GenBank mRNA | FM051934.1 | - | 1 | 209 | ||
FM055064.1 | - | 1 | 183 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Srsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109552.2 | NP_001103022.1 | 1 | 230 | ||
GenBank mRNA | FM051934.1 | - | 1 | 209 | ||
FM055064.1 | - | 1 | 183 |