siRNA Details
| Gene Symbol: Rac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768496.1 | XP_008766718.1 | 5 | 961 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rac3| Gene Symbol: Rac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768496.1 | XP_008766718.1 | 5 | 961 | ||