siRNA Details
| Gene Symbol: Foxk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107075.1 | NP_001100545.1 | 1, 2 | 125 | ||
| XM_006247890.2 | XP_006247952.1 | 1, 2 | 522 | |||
| XM_006247891.2 | XP_006247953.1 | 1, 2 | 522 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxk2| Gene Symbol: Foxk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107075.1 | NP_001100545.1 | 1, 2 | 125 | ||
| XM_006247890.2 | XP_006247952.1 | 1, 2 | 522 | |||
| XM_006247891.2 | XP_006247953.1 | 1, 2 | 522 | |||