siRNA Details
| Gene Symbol: Mab21l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109497.1 | NP_001102967.1 | 1 | 1219 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mab21l1| Gene Symbol: Mab21l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109497.1 | NP_001102967.1 | 1 | 1219 | ||