siRNA Details
Gene Symbol: RGD1564963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242289.2 | XP_006242351.1 | 1 | 377 |
Gene Symbol: LOC100362967 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233349.2 | XP_006233411.1 | 1 | 248 |
Gene Symbol: LOC100912132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252449.2 | XP_006252511.2 | 1 | 240 |
Gene Symbol: RGD1563861 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_574109.5 | XP_574109.1 | 1 | 607 |
Gene Symbol: Rpl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031100.1 | NP_112362.1 | 7 | 622 | ||
GenBank mRNA | BC058467.1 | - | 7 | 611 | 6918973 | |
FQ210278.1 | - | 7 | 606 | |||
FQ210394.1 | - | 3 | 324 | |||
FQ212456.1 | - | 3 | 324 | |||
FQ217196.1 | - | 3 | 324 | |||
FQ217560.1 | - | 3 | 324 | |||
FQ217981.1 | - | 3 | 324 | |||
FQ218098.1 | - | 7 | 606 | |||
FQ221411.1 | - | 3 | 324 | |||
FQ221714.1 | - | 3 | 324 | |||
FQ221864.1 | - | 7 | 606 | |||
FQ222021.1 | - | 7 | 606 | |||
FQ222201.1 | - | 3 | 324 | |||
FQ222372.1 | - | 3 | 324 | |||
FQ222670.1 | - | 3 | 324 | |||
FQ222782.1 | - | 3 | 324 | |||
FQ222847.1 | - | 3 | 324 | |||
FQ223155.1 | - | 3 | 324 | |||
FQ223157.1 | - | 3 | 324 | |||
FQ224407.1 | - | 3 | 324 | |||
FQ224785.1 | - | 3 | 324 | |||
FQ226301.1 | - | 3 | 324 | |||
FQ228167.1 | - | 3 | 324 | |||
FQ229032.1 | - | 7 | 605 | |||
FQ229696.1 | - | 5 | 733 | |||
FQ231133.1 | - | 3 | 324 | |||
FQ231791.1 | - | 3 | 324 | |||
FQ233952.1 | - | 7 | 606 | |||
FQ235226.1 | - | 3 | 324 | |||
X87106.1 | - | 7 | 622 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
