siRNA Details
| Gene Symbol: Upk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109523.1 | NP_001102993.1 | 5 | 684 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Upk2| Gene Symbol: Upk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109523.1 | NP_001102993.1 | 5 | 684 | ||