siRNA Details
| Gene Symbol: Smim5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163002.1 | NP_001156474.1 | 3 | 753 | ||
| NM_001277479.1 | NP_001264408.1 | 4 | 911 | |||
| XM_006247731.1 | XP_006247793.1 | 2 | 6974 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smim5| Gene Symbol: Smim5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163002.1 | NP_001156474.1 | 3 | 753 | ||
| NM_001277479.1 | NP_001264408.1 | 4 | 911 | |||
| XM_006247731.1 | XP_006247793.1 | 2 | 6974 | |||