siRNA Details
| Gene Symbol: Syce3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135253.1 | NP_001128725.1 | 2 | 582 | ||
| XM_006242244.2 | XP_006242306.1 | 3 | 857 | |||
| XM_006242246.2 | XP_006242308.1 | 3 | 732 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syce3| Gene Symbol: Syce3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135253.1 | NP_001128725.1 | 2 | 582 | ||
| XM_006242244.2 | XP_006242306.1 | 3 | 857 | |||
| XM_006242246.2 | XP_006242308.1 | 3 | 732 | |||