siRNA Details
| Gene Symbol: POU6F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166018.1 | NP_001159490.1 | 6, 7 | 1036 | ||
| NM_007252.3 | NP_009183.3 | 6, 7 | 1036 | |||
| XM_011515113.2 | XP_011513415.1 | 5, 6 | 1056 | |||
| GenBank mRNA | U91935.1 | - | 5, 6 | 979 | ||

POU6F2
Genomic Map