siRNA Details
Gene Symbol: Cyp4f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173124.1 | NP_775147.1 | 2324 | |||
XM_006241051.2 | XP_006241113.1 | 13 | 2393 | |||
XM_008765157.1 | XP_008763379.1 | 12 | 2341 | |||
GenBank mRNA | BC129085.1 | - | 13 | 2285 | 7375658 | |
U39207.1 | - | 2324 |
Gene Symbol: LOC100912471 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241096.2 | XP_006241158.1 | 3218 | |||
XM_006241097.2 | XP_006241159.1 | 11 | 1960 |
Gene Symbol: LOC691320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241044.2 | XP_006241106.1 | 8 | 3218 | ||
XM_006241045.1 | XP_006241107.1 | 10 | 1927 |
