siRNA Details
Gene Symbol: LOC100912557 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FQ211017.1 | - | 4 | 320 | ||
FQ217325.1 | - | 3 | 274 | |||
FQ222783.1 | - | 4 | 323 | |||
FQ224630.1 | - | 4 | 302 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100912557 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FQ211017.1 | - | 4 | 320 | ||
FQ217325.1 | - | 3 | 274 | |||
FQ222783.1 | - | 4 | 323 | |||
FQ224630.1 | - | 4 | 302 |