siRNA Details
| Gene Symbol: LOC100912557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | FQ211017.1 | - | 4 | 248 | ||
| FQ217325.1 | - | 3 | 202 | |||
| FQ222783.1 | - | 4 | 251 | |||
| FQ224630.1 | - | 4 | 230 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100912557| Gene Symbol: LOC100912557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | FQ211017.1 | - | 4 | 248 | ||
| FQ217325.1 | - | 3 | 202 | |||
| FQ222783.1 | - | 4 | 251 | |||
| FQ224630.1 | - | 4 | 230 | |||