siRNA Details
| Gene Symbol: Gid4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750840.3 | XP_003750888.1 | 1 | 649 | ||
| GenBank mRNA | BC169088.1 | - | 1 | 169 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gid4| Gene Symbol: Gid4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750840.3 | XP_003750888.1 | 1 | 649 | ||
| GenBank mRNA | BC169088.1 | - | 1 | 169 | ||