siRNA Details
Gene Symbol: Mlip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766417.1 | XP_008764639.1 | 2 | 522 | ||
XM_008766420.1 | XP_008764642.1 | 2 | 255 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mlip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766417.1 | XP_008764639.1 | 2 | 522 | ||
XM_008766420.1 | XP_008764642.1 | 2 | 255 |