siRNA Details
| Gene Symbol: Hmgcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024386.1 | NP_077362.1 | 9 | 949 | ||
| GenBank mRNA | BC061797.1 | - | 9 | 951 | 5598033 | |
| FQ209392.1 | - | 9 | 948 | |||
| FQ209616.1 | - | 9 | 953 | |||
| FQ234382.1 | - | 9 | 947 | |||
| Y10054.1 | - | 9 | 949 | |||

Hmgcl
Genomic Map