siRNA Details
Gene Symbol: Opn1sw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031015.1 | NP_112277.1 | 1 | 225 | ||
GenBank mRNA | AF051163.1 | - | 1 | 177 | ||
U63972.1 | - | 1 | 225 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
