siRNA Details
| Gene Symbol: Ocln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031329.2 | NP_112619.2 | 7 | 1471 | ||
| XM_006231853.2 | XP_006231915.1 | 7 | 1467 | |||
| XM_006231854.2 | XP_006231916.1 | 7 | 1511 | |||
| XM_006231855.2 | XP_006231917.1 | 6 | 1391 | |||
| GenBank mRNA | BC062037.1 | - | 7 | 1471 | 5623744 | |

Ocln
Genomic Map