siRNA Details
| Gene Symbol: Fads2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031344.2 | NP_112634.1 | 9 | 1211 | ||
| XM_008760244.1 | XP_008758466.1 | 9 | 2720 | |||
| GenBank mRNA | AB021980.1 | - | 9 | 1141 | ||
| BC081776.1 | - | 9 | 1211 | 7128630 | ||
| FQ210893.1 | - | 9 | 1140 | |||
| HQ909027.1 | - | 9 | 1045 | |||

Fads2
Genomic Map