siRNA Details
| Gene Symbol: Pex3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031350.1 | NP_112640.1 | 10 | 934 | ||
| GenBank mRNA | AB035306.1 | - | 10 | 934 | ||
| BC062046.1 | - | 10 | 1025 | 5597999 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pex3| Gene Symbol: Pex3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031350.1 | NP_112640.1 | 10 | 934 | ||
| GenBank mRNA | AB035306.1 | - | 10 | 934 | ||
| BC062046.1 | - | 10 | 1025 | 5597999 | ||