siRNA Details
| Gene Symbol: Tpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031357.1 | NP_112647.1 | 11 | 1322 | ||
| GenBank mRNA | AB043870.1 | - | 11 | 1322 | ||
| AY724484.1 | - | 11 | 1323 | |||
| BC081775.1 | - | 11 | 1331 | 7107183 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpp1| Gene Symbol: Tpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031357.1 | NP_112647.1 | 11 | 1322 | ||
| GenBank mRNA | AB043870.1 | - | 11 | 1322 | ||
| AY724484.1 | - | 11 | 1323 | |||
| BC081775.1 | - | 11 | 1331 | 7107183 | ||