siRNA Details
| Gene Symbol: CYP4F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007253.3 | NP_009184.1 | 11 | 1334 | ||
| GenBank mRNA | AF133298.1 | - | 10 | 1277 | ||
| AK092612.1 | - | 1 | 1381 | |||
| AK096809.1 | - | 7 | 2988 | |||
| AK300539.1 | - | 7 | 885 | |||
| AK314685.1 | - | 11 | 1332 | |||

CYP4F8
Genomic Map