siRNA Details
| Gene Symbol: Gipc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053341.1 | NP_445793.1 | 6 | 988 | ||
| XM_006255311.2 | XP_006255373.1 | 6 | 3253 | |||
| XM_006255312.2 | XP_006255374.1 | 7 | 2247 | |||
| XM_008772420.1 | XP_008770642.1 | 7 | 2177 | |||
| GenBank mRNA | AF032120.1 | - | 6 | 988 | ||
| BC061964.1 | - | 5 | 1796 | |||
| BC070505.1 | - | 7 | 1031 | 7097697 | ||

Gipc1
Genomic Map