siRNA Details
| Gene Symbol: Cnga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053497.1 | NP_445949.1 | 8 | 1389 | ||
| GenBank mRNA | U70257.1 | - | 1 | 114 | ||
| U93851.1 | - | 8 | 1389 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnga1| Gene Symbol: Cnga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053497.1 | NP_445949.1 | 8 | 1389 | ||
| GenBank mRNA | U70257.1 | - | 1 | 114 | ||
| U93851.1 | - | 8 | 1389 | |||