siRNA Details
| Gene Symbol: Shc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164060.1 | NP_001157532.1 | 12 | 1847 | ||
| NM_053517.2 | NP_445969.2 | 13 | 1414 | |||
| GenBank mRNA | BC088298.1 | - | 13 | 1414 | 7323045 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shc1| Gene Symbol: Shc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164060.1 | NP_001157532.1 | 12 | 1847 | ||
| NM_053517.2 | NP_445969.2 | 13 | 1414 | |||
| GenBank mRNA | BC088298.1 | - | 13 | 1414 | 7323045 | |