siRNA Details
| Gene Symbol: Sfrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053544.1 | NP_445996.1 | 1, 2 | 547 | ||
| GenBank mRNA | AF012891.1 | - | 1, 2 | 547 | ||
| AF140346.1 | - | 1, 2 | 698 | |||
| AF220608.1 | - | 795 | ||||
More Information
Other Species Targeted
Set Membership: |
|

Sfrp4
Genomic Map