siRNA Details
| Gene Symbol: Sfrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053544.1 | NP_445996.1 | 4 | 746 | ||
| GenBank mRNA | AF012891.1 | - | 4 | 746 | ||
| AF140346.1 | - | 4 | 897 | |||
| AF220608.1 | - | 994 | ||||
| FQ220237.1 | - | 1 | 55 | |||
| FQ229078.1 | - | 1 | 68 | |||
| FQ229224.1 | - | 3 | 177 | |||
More Information
Set Membership: |
|

Sfrp4
Genomic Map