siRNA Details
| Gene Symbol: Bcl2a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007534.3 | NP_031560.2 | 2 | 530 | ||
| GenBank mRNA | AK138369.1 | - | 2 | 510 | ||
| AK152200.1 | - | 2 | 500 | |||
| BC100462.1 | - | 2 | 507 | 30295442 | ||
| BC116241.1 | - | 2 | 530 | 40091848 | ||
| BP773251.1 | - | 2 | 88 | |||
| Gene Symbol: Bcl2a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009742.3 | NP_033872.1 | 2 | 574 | ||
| GenBank mRNA | BC028762.1 | - | 520 | 1226745 | ||
| CT010250.1 | - | 443 | ||||
| L16462.1 | - | 2 | 574 | |||
| Gene Symbol: Bcl2a1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007536.2 | NP_031562.1 | 2 | 574 | ||
| GenBank mRNA | AK165576.1 | - | 2 | 326 | ||
| BC027536.1 | - | 2 | 487 | 1363928 | ||
| BC053965.1 | - | 2 | 596 | |||
| BC116240.2 | - | 2 | 530 | 40091847 | ||

Bcl2a1b
Genomic Map