siRNA Details
| Gene Symbol: Gm9112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177365.1 | NP_001170836.1 | 2 | 289 | ||
| GenBank mRNA | CJ042093.1 | - | 2 | 290 | ||
| CJ042401.1 | - | 2 | 255 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9112| Gene Symbol: Gm9112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177365.1 | NP_001170836.1 | 2 | 289 | ||
| GenBank mRNA | CJ042093.1 | - | 2 | 290 | ||
| CJ042401.1 | - | 2 | 255 | |||