siRNA Details
| Gene Symbol: SLU7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006425.4 | NP_006416.3 | 7, 8 | 850 | ||
| XM_005265793.3 | XP_005265850.1 | 7, 8 | 1197 | |||
| XM_011534418.2 | XP_011532720.1 | 8, 9 | 900 | |||
| GenBank mRNA | AK314564.1 | - | 7, 8 | 770 | ||
| CA976106.1 | - | 5, 6 | 441 | |||

SLU7
Genomic Map