siRNA Details
| Gene Symbol: KERA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007035.3 | NP_008966.1 | 2 | 916 | ||
| GenBank mRNA | AF063301.1 | - | 1 | 305 | ||
| AF205403.1 | - | 2 | 545 | |||
| AK314990.1 | - | 2 | 478 | |||
| BC032667.2 | - | 1 | 306 | 5541591 | ||
| BM722433.1 | - | 2 | 449 | |||
| CV571245.1 | - | 2 | 463 | |||

KERA
Genomic Map