siRNA Details
| Gene Symbol: CIT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206999.1 | NP_001193928.1 | 20 | 2458 | ||
| NM_007174.2 | NP_009105.1 | 19 | 2332 | |||
| XM_006719206.2 | XP_006719269.1 | 20 | 2485 | |||
| XM_011537783.1 | XP_011536085.1 | 20 | 2485 | |||
| XM_011537784.1 | XP_011536086.1 | 20 | 2485 | |||
| XM_011537785.1 | XP_011536087.1 | 20 | 2485 | |||
| XM_011537787.1 | XP_011536089.1 | 19 | 2359 | |||
| XM_011537788.1 | XP_011536090.1 | 19 | 2359 | |||
| XM_011537789.1 | XP_011536091.1 | 12 | 1620 | |||
| XM_011537790.2 | XP_011536092.1 | 11 | 1188 | |||
| XM_011537791.2 | XP_011536093.1 | 11 | 2173 | |||
| XM_011537792.2 | XP_011536094.1 | 11 | 2318 | |||
| XM_017018735.1 | XP_016874224.1 | 20 | 2485 | |||
| XM_017018736.1 | XP_016874225.1 | 20 | 2485 | |||
| XM_017018737.1 | XP_016874226.1 | 20 | 2485 | |||
| XM_017018738.1 | XP_016874227.1 | 9 | 1729 | |||
| XM_017018739.1 | XP_016874228.1 | 10 | 2594 | |||
| GenBank mRNA | AB023166.2 | - | 11 | 2318 | ||
| AY257469.1 | - | 19 | 2329 | |||
| AY681966.1 | - | 20 | 2458 | |||

CIT
Genomic Map