siRNA Details
| Gene Symbol: MGAT4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160154.1 | NP_001153626.1 | 8 | 825 | ||
| NM_012214.2 | NP_036346.1 | 11 | 1409 | |||
| GenBank mRNA | AB000616.1 | - | 10 | 1230 | ||
| AK291533.1 | - | 10 | 1219 | |||
| AK304393.1 | - | 8 | 825 | |||
| AK314024.1 | - | 11 | 1369 | |||
| BC127107.1 | - | 10 | 1307 | 40124731 | ||
| BC127108.1 | - | 10 | 1307 | 40124741 | ||

MGAT4A
Genomic Map