siRNA Details
| Gene Symbol: SLC2A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145099.1 | NP_001138571.1 | 9 | 2312 | ||
| NM_017585.3 | NP_060055.2 | 10 | 2498 | |||
| XM_011518189.2 | XP_011516491.2 | 10 | 2475 | |||
| XM_017014237.1 | XP_016869726.1 | 10 | 2466 | |||
| XM_017014238.1 | XP_016869727.1 | 10 | 2470 | |||
| GenBank mRNA | AJ011372.2 | - | 10 | 2440 | ||

SLC2A6
Genomic Map