siRNA Details
Gene Symbol: NISCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007184.3 | NP_009115.2 | 21 | 4075 | ||
XM_005264839.2 | XP_005264896.1 | 20 | 3924 | |||
XM_006712955.2 | XP_006713018.1 | 9 | 4602 | |||
XM_017005643.1 | XP_016861132.1 | 12 | 2916 | |||
GenBank mRNA | AB023192.1 | - | 22 | 4013 | ||
AF082516.1 | - | 21 | 3967 | |||
AK027331.1 | - | 6 | 1976 | |||
AK027428.1 | - | 21 | 3953 | |||
AK075000.1 | - | 6 | 1852 | |||
AK097732.1 | - | 5 | 1038 | |||
AK291398.1 | - | 21 | 3979 | |||
AK297657.1 | - | 7 | 751 | |||
AK302441.1 | - | 12 | 2963 | |||
AK315001.1 | - | 21 | 3979 | |||
AL117432.1 | - | 8 | 2534 | |||
BC038102.2 | - | 21 | 3979 | 5268247 | ||
BC054494.1 | - | 21 | 3973 | 6195532 | ||
BC056900.1 | - | 21 | 3945 | 6143316 |

More Information
Panther Classification:
Molecular Function -



Gene Ontology Categories:
