siRNA Details
| Gene Symbol: CARD16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017534.1 | NP_001017534.1 | 3 | 431 | ||
| NM_052889.2 | NP_443121.1 | 4 | 494 | |||
| XM_011542583.2 | XP_011540885.1 | 3 | 623 | |||
| GenBank mRNA | AK125640.1 | - | 2 | 3015 | ||
| AY885669.1 | - | 3 | 422 | |||
| BC070196.1 | - | 4 | 494 | |||
| BC104635.1 | - | 4 | 498 | |||

CARD16
Genomic Map