siRNA Details
Gene Symbol: MRGPRE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039165.2 | NP_001034254.2 | 2 | 1109 | ||
GenBank mRNA | AK293940.1 | - | 2 | 1109 | ||
BC104889.1 | - | 1 | 833 | 8143892 | ||
BC112203.1 | - | 1 | 845 | 8327671 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
