siRNA Details
| Gene Symbol: ACSM2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308169.1 | NP_001295098.1 | 13 | 1720 | ||
| NM_001308172.1 | NP_001295101.1 | 14 | 1905 | |||
| NM_001308954.1 | NP_001295883.1 | 15 | 2287 | |||
| XM_017022923.1 | XP_016878412.1 | 15 | 1880 | |||
| XM_017022926.1 | XP_016878415.1 | 12 | 1422 | |||
| GenBank mRNA | AK055867.1 | - | 15 | 1701 | ||
| AK091878.1 | - | 7 | 1871 | |||
| AK096039.1 | - | 15 | 2146 | |||
| AK298359.1 | - | 13 | 1609 | |||
| BC125176.1 | - | 14 | 1742 | 40121949 | ||

ACSM2A
Genomic Map