siRNA Details
| Gene Symbol: COX6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004373.3 | NP_004364.2 | 3 | 446 | ||
| GenBank mRNA | BC007723.2 | - | 1 | 411 | 4300921 | |
| BC070186.1 | - | 1 | 419 | 6654310 | ||
| BC107861.1 | - | 1 | 427 | 4664958 | ||
| BP378295.1 | - | 3 | 417 | |||
| HE647860.1 | - | 3 | 409 | |||
| X15341.1 | - | 3 | 431 | |||

COX6A1
Genomic Map