siRNA Details
Gene Symbol: TAAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138327.2 | NP_612200.1 | 1 | 924 | ||
GenBank mRNA | AY180374.1 | - | 1 | 901 | ||
BC101825.1 | - | 1 | 924 | 8069331 | ||
BC112194.1 | - | 1 | 936 | 8327662 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
