siRNA Details
| Gene Symbol: TRIM40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286633.1 | NP_001273562.1 | 2 | 624 | ||
| NM_138700.4 | NP_619645.1 | 1 | 542 | |||
| XM_011514305.1 | XP_011512607.1 | 2 | 1055 | |||
| XM_011514306.1 | XP_011512608.1 | 3 | 921 | |||
| XM_011514308.1 | XP_011512610.1 | 2 | 1055 | |||
| XM_011514309.1 | XP_011512611.1 | 2 | 1055 | |||
| GenBank mRNA | AF489517.1 | - | 1 | 265 | ||
| BC060785.1 | - | 2 | 624 | 30340682 | ||

TRIM40
Genomic Map