siRNA Details
| Gene Symbol: CPM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005502.2 | NP_001005502.1 | 4 | 366 | ||
| NM_001874.4 | NP_001865.1 | 4 | 398 | |||
| NM_198320.3 | NP_938079.1 | 4 | 384 | |||
| GenBank mRNA | AF368463.1 | - | 4 | 356 | ||
| AK313180.1 | - | 4 | 356 | |||
| BC022276.1 | - | 4 | 356 | 4768771 | ||
| CB959900.1 | - | 383 | ||||
| CB989009.1 | - | 4 | 454 | |||
| CD109360.1 | - | 4 | 383 | |||
| DA205368.1 | - | 4 | 385 | |||
| DB006895.1 | - | 4 | 369 | |||
| J04970.2 | - | 4 | 398 | |||

CPM
Genomic Map