siRNA Details
| Gene Symbol: RAB40A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080879.2 | NP_543155.2 | 3 | 885 | ||
| XM_005262083.4 | XP_005262140.1 | 2 | 1362 | |||
| XM_011530872.2 | XP_011529174.1 | 2 | 1359 | |||
| GenBank mRNA | AB232637.1 | - | 1 | 543 | ||
| AF132748.1 | - | 1 | 608 | |||
| AF422143.1 | - | 3 | 885 | |||
| AK056859.1 | - | 1 | 2662 | |||
| BC074854.2 | - | 1 | 574 | 30915461 | ||
| BC074855.2 | - | 1 | 574 | 30915224 | ||
| BC113501.1 | - | 1 | 626 | 8322553 | ||
| BC117232.1 | - | 1 | 626 | 40125783 | ||

RAB40A
Genomic Map