siRNA Details
| Gene Symbol: CSTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005213.3 | NP_005204.1 | 1, 2 | 196 | ||
| GenBank mRNA | AK291308.1 | - | 1, 2 | 112 | ||
| AV716538.1 | - | 1, 2 | 210 | |||
| BC010379.1 | - | 1, 2 | 116 | 4063525 | ||
| CR456914.1 | - | 1, 2 | 67 | |||
| X05978.1 | - | 1, 2 | 112 | |||

CSTA
Genomic Map