siRNA Details
| Gene Symbol: ZNF563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145276.2 | NP_660319.1 | 3 | 386 | ||
| XM_005259750.4 | XP_005259807.1 | 3 | 385 | |||
| XM_005259751.3 | XP_005259808.1 | 3 | 386 | |||
| XM_006722651.3 | XP_006722714.1 | 3 | 384 | |||
| XM_011527698.1 | XP_011526000.1 | 3 | 206 | |||
| XM_017026332.1 | XP_016881821.1 | 2 | 122 | |||
| GenBank mRNA | AK092116.1 | - | 3 | 386 | ||
| AK313755.1 | - | 3 | 290 | |||
| BC022523.1 | - | 3 | 354 | 4794529 | ||
| DA687547.1 | - | 3 | 386 | |||

ZNF563
Genomic Map