siRNA Details
| Gene Symbol: PPTC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139283.1 | NP_644812.1 | 4 | 920 | ||
| GenBank mRNA | AF385435.1 | - | 4 | 920 | ||
| AK124744.1 | - | 5 | 764 | |||
| AY357944.1 | - | 4 | 689 | |||
| BC111551.1 | - | 5 | 722 | 40009728 | ||
| CR749216.1 | - | 1 | 42 | |||

PPTC7
Genomic Map