siRNA Details
| Gene Symbol: DMPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081560.2 | NP_001075029.1 | 7 | 1006 | ||
| NM_001081562.2 | NP_001075031.1 | 7 | 1006 | |||
| NM_001081563.2 | NP_001075032.1 | 6 | 1375 | |||
| NM_001288764.1 | NP_001275693.1 | 8 | 1065 | |||
| NM_001288765.1 | NP_001275694.1 | 5 | 646 | |||
| NM_001288766.1 | NP_001275695.1 | 7 | 1006 | |||
| NM_004409.4 | NP_004400.4 | 7 | 1006 | |||
| GenBank mRNA | AB209363.1 | - | 7 | 1849 | ||
| AK297299.1 | - | 5 | 597 | |||
| AK302874.1 | - | 7 | 939 | |||
| BC026328.1 | - | 8 | 1060 | 4792104 | ||
| BC062553.1 | - | 7 | 927 | 6480902 | ||
| KJ534826.1 | - | 6 | 831 | |||
| KJ534827.1 | - | 6 | 831 | |||
| KJ534828.1 | - | 6 | 831 | |||
| L19268.1 | - | 7 | 928 | |||
| M87312.1 | - | 6 | 1376 | |||
| M94203.1 | - | 7 | 678 | |||
| S72883.1 | - | 7 | 837 | |||
 Back To Top
 Back To Top
 DMPK
DMPK Genomic Map
Genomic Map Back To Top
 Back To Top