siRNA Details
| Gene Symbol: ACOT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007274.3 | NP_009205.3 | 3 | 762 | ||
| NM_181864.2 | NP_863654.1 | 3 | 582 | |||
| NM_181865.2 | NP_863655.1 | 3 | 387 | |||
| NM_181866.2 | NP_863656.1 | 3 | 371 | |||
| GenBank mRNA | AB074415.1 | - | 3 | 357 | ||
| AB074416.1 | - | 3 | 357 | |||
| AB074417.1 | - | 3 | 572 | |||
| AB074418.1 | - | 3 | 343 | |||
| AB074419.1 | - | 3 | 334 | |||
| AK057168.1 | - | 3 | 758 | |||
| AK289572.1 | - | 3 | 373 | |||
| AK290097.1 | - | 3 | 437 | |||
| AK291583.1 | - | 3 | 356 | |||
| AK292202.1 | - | 3 | 385 | |||
| AK300831.1 | - | 2 | 252 | |||
| AK304465.1 | - | 3 | 389 | |||
| BC017365.2 | - | 3 | 711 | 3507488 | ||
| BM807217.1 | - | 3 | 574 | |||
| BQ067921.1 | - | 430 | ||||
| BT006888.1 | - | 3 | 299 | |||
| BU594651.1 | - | 3 | 393 | |||
| D88894.2 | - | 3 | 357 | |||
| U91316.1 | - | 3 | 448 | |||

ACOT7
Genomic Map