siRNA Details
| Gene Symbol: ENO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193503.1 | NP_001180432.1 | 6 | 742 | ||
| NM_001976.4 | NP_001967.3 | 8 | 913 | |||
| NM_053013.3 | NP_443739.3 | 8 | 871 | |||
| XM_005256521.2 | XP_005256578.1 | 8 | 1299 | |||
| XM_011523729.1 | XP_011522031.1 | 8 | 889 | |||
| XM_017024346.1 | XP_016879835.1 | 8 | 1306 | |||
| GenBank mRNA | AK300662.1 | - | 8 | 757 | ||
| AK300673.1 | - | 7 | 733 | |||
| AK300709.1 | - | 6 | 712 | |||
| AK309864.1 | - | 9 | 964 | |||
| BC017249.2 | - | 8 | 828 | 4902149 | ||
| X16504.1 | - | 7 | 775 | |||
| X51957.1 | - | 7 | 787 | |||

ENO3
Genomic Map