siRNA Details
| Gene Symbol: TUSC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007275.2 | NP_009206.1 | 1, 2 | 290 | ||
| GenBank mRNA | AF055479.1 | - | 1, 2 | 298 | ||
| AK124058.1 | - | 2, 3 | 224 | |||
| AK311998.1 | - | 1, 2 | 280 | |||
| BC023976.2 | - | 1, 2 | 242 | 3504783 | ||
| CA392689.1 | - | 1, 2 | 290 | |||

TUSC2
Genomic Map