siRNA Details
| Gene Symbol: GGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015044.4 | NP_055859.1 | 2 | 209 | ||
| XM_017023074.1 | XP_016878563.1 | 3 | 3898 | |||
| XM_017023075.1 | XP_016878564.1 | 2 | 207 | |||
| GenBank mRNA | AF165531.1 | - | 2 | 147 | ||
| AF190863.1 | - | 2 | 153 | |||
| AF233522.1 | - | 2 | 152 | |||
| AF323754.1 | - | 2 | 151 | |||
| AK024619.1 | - | 2 | 209 | |||
| AK308425.1 | - | 2 | 196 | |||
| BC000284.1 | - | 2 | 191 | 3356490 | ||
| CN423115.1 | - | 2 | 206 | |||
| DA439738.1 | - | 2 | 209 | |||

GGA2
Genomic Map